Thermo Scientific Chemicals

Solketal, 97%, Thermo Scientific Chemicals

Catalog number: L02814.36
500 g, Each
Thermo Scientific Chemicals

Solketal, 97%, Thermo Scientific Chemicals

Catalog number: L02814.36
500 g, Each
Quantity
Catalog number: L02814.36
also known as L02814-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-79-8
IUPAC Name
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Molecular Formula
C6H12O3
InChI Key
RNVYQYLELCKWAN-UHFFFAOYNA-N
SMILES
CC1(C)OCC(CO)O1
Assay (GC)
≥96.0%
Refractive Index
1.4320-1.4370 @ 20?C
Appearance (Color)
Clear, colorless
Form
Liquid

Description

Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .

Solubility
Soluble in water (fully), alcohol, acetals, esters, and ethers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are acids, Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text