1,3-Diphenylacetone, 98+%, Thermo Scientific Chemicals
1,3-Diphenylacetone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diphenylacetone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L02830.22
also known as L02830-22
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
Add to cart
1,3-Diphenylacetone, 98+%, Thermo Scientific Chemicals
Catalog numberL02830.22
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS102-04-5
IUPAC Name1,3-diphenylpropan-2-one
Molecular FormulaC15H14O
InChI KeyYFKBXYGUSOXJGS-UHFFFAOYSA-N
SMILESO=C(CC1=CC=CC=C1)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystalline or fused solid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)30-36?C
1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylacetone is used in the aldol condensation reaction with benzil (a dicarbonyl) and base to create tetraphenylcyclopentadienone.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hoang, G. T.; Kubo, T.; Young, V. G.; Kautzky, J. A.; Wissinger, J. E. Illustrating the Utility of X-ray Crystallography for Structure Elucidation through a Tandem Aldol Condensation/Diels-Alder Reaction Sequence. J. Chem. Educ. 2015, 92 (8), 1381-1384.
  2. Cheung, K. Y.; Xu, X.; Miao, Q. Aromatic Saddles Containing Two Heptagons. J. Am. Chem. Soc. 2015, 137 (11), 3910-3914.