Thermo Scientific Chemicals

5-Chloro-1-pentyne, 98%, Thermo Scientific Chemicals

Catalog number: L02835.14
25 g, Each
Thermo Scientific Chemicals

5-Chloro-1-pentyne, 98%, Thermo Scientific Chemicals

Catalog number: L02835.14
25 g, Each
Quantity
Catalog number: L02835.14
also known as L02835-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14267-92-6
IUPAC Name
5-chloropent-1-yne
Molecular Formula
C5H7Cl
InChI Key
UXFIKVWAAMKFQE-UHFFFAOYSA-N
SMILES
ClCCCC#C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4425-1.4465 @ 20?C

Description

5-Chloro-1-pentyne is used with α-diazoesters in a copper-catalyzed synthesis of pyrazoles. It is also used in the synthesis of 5-chloro-2-trimethylstannyl-1-pentene by reacting with Me3SnCu.Me2S.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-1-pentyne is used with α-diazoesters in a copper-catalyzed synthesis of pyrazoles. It is also used in the synthesis of 5-chloro-2-trimethylstannyl-1-pentene by reacting with Me3SnCu.Me2S.

Solubility
Not miscible in water.

Notes
Store at 4°C. Protect from heat. Incompatible with heavy metal powders, heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text