Thermo Scientific Chemicals

Cholesteryl nonanoate, Thermo Scientific Chemicals

Catalog number: L02857.22
100 g, Each
Thermo Scientific Chemicals

Cholesteryl nonanoate, Thermo Scientific Chemicals

Catalog number: L02857.22
100 g, Each
Quantity
Catalog number: L02857.22
also known as L02857-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1182-66-7
IUPAC Name
9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl nonanoate
Molecular Formula
C36H62O2
InChI Key
WCLNGBQPTVENHV-UHFFFAOYNA-N
SMILES
CCCCCCCCC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C
Appearance (Color)
White
Melting Point
75-79?C (≤2.5?C range)
Optical Rotation
-30 ? 1? (c=5 in chloroform)
Form
powder

Description

Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

Solubility
Soluble in Chloroform, Tetrahydrofuran(THF). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text