Thermo Scientific Chemicals

Phenyltrimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: L02876.14
25 g, Each
Thermo Scientific Chemicals

Phenyltrimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: L02876.14
25 g, Each
Quantity
Catalog number: L02876.14
also known as L02876-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
768-32-1
IUPAC Name
trimethyl(phenyl)silane
Molecular Formula
C9H14Si
InChI Key
KXFSUVJPEQYUGN-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4885-1.4925 @ 20°C

Description

It is employed as intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical and organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text