Thermo Scientific Chemicals

1-Undecyne, 97%, Thermo Scientific Chemicals

Catalog number: L02883.09
10 g, Each
Thermo Scientific Chemicals

1-Undecyne, 97%, Thermo Scientific Chemicals

Catalog number: L02883.09
10 g, Each
Quantity
Catalog number: L02883.09
also known as L02883-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2243-98-3
IUPAC Name
undec-1-yne
Molecular Formula
C11H20
InChI Key
YVSFLVNWJIEJRV-UHFFFAOYSA-N
SMILES
CCCCCCCCCC#C
Assay (GC)
≥96.0% (UK-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Refractive Index
1.4285-1.4350 @ 20?C (UK-sourced material)
Form
Liquid
Assay from Supplier's CofA
≥96.0% (US-sourced material)

Description

1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

Solubility
Miscible with acetone, ether, ethanol and benzene. Slightly miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text