Thermo Scientific Chemicals

Methyl oleate, tech. C18 71-90%, C18:1 >65% of C18, Thermo Scientific Chemicals

Catalog number: L02937.AU
1000 mL, Each
Thermo Scientific Chemicals

Methyl oleate, tech. C18 71-90%, C18:1 >65% of C18, Thermo Scientific Chemicals

Catalog number: L02937.AU
1000 mL, Each
Quantity
Catalog number: L02937.AU
also known as L02937-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-62-9
IUPAC Name
methyl (9E)-octadec-9-enoate
Molecular Formula
C19H36O2
InChI Key
QYDYPVFESGNLHU-ZHACJKMWSA-N
SMILES
CCCCCCCCC=CCCCCCCCC(=O)OC
Appearance (Color)
Clear or slightly turbid, colorless to pale yellow
Acid Value
≤1.0mg KOH/g
Form
Liquid
Iodine value
80-97 gI₂/100g
Comment
C18:1 - ≥65.0%

Description

Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers, for duplicating ink, rubber and wax. It finds application as lubricant and lubricant additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers, for duplicating ink, rubber and wax. It finds application as lubricant and lubricant additive.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text