Thermo Scientific Chemicals

Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals

Catalog number: L02940.14
25 g, Each
Thermo Scientific Chemicals

Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals

Catalog number: L02940.14
25 g, Each
Quantity
Catalog number: L02940.14
also known as L02940-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-21-0
IUPAC Name
4,10-dioxapentacyclo[6.3.1.0²,⁷.0³,⁵.0⁹,¹¹]dodecane
Molecular Formula
C10H12O2
InChI Key
BQQUFAMSJAKLNB-UHFFFAOYNA-N
SMILES
C1C2OC2C2C3CC(C4OC34)C12
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or lumps
Melting Point (clear melt)
175-192?C
Assay (GC)
≥97.5%

Description

Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text