Thermo Scientific Chemicals

1-Dodecyne, 97%, Thermo Scientific Chemicals

Catalog number: L02977.14
25 g, Each
Thermo Scientific Chemicals

1-Dodecyne, 97%, Thermo Scientific Chemicals

Catalog number: L02977.14
25 g, Each
Quantity
Catalog number: L02977.14
also known as L02977-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
765-03-7
IUPAC Name
dodec-1-yne
Molecular Formula
C12H22
InChI Key
ZVDBUOGYYYNMQI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC#C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms (non-U.S. specification)
Refractive Index
1.4320-1.4370 @ 20°C (non-U.S. specification)
Form
Liquid

Description

1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Solubility
Not miscible in water.

Notes
Incompatible with heavy metal powders and oxidizing agents. Store between 2 - 8°C. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text