Thermo Scientific Chemicals

3-(3,4-Dichlorophenyl)-1,1-dimethylurea, 97%, Thermo Scientific Chemicals

Catalog number: L02986.18
50 g, Each
Thermo Scientific Chemicals

3-(3,4-Dichlorophenyl)-1,1-dimethylurea, 97%, Thermo Scientific Chemicals

Catalog number: L02986.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L02986.18
also known as L02986-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
330-54-1
IUPAC Name
1-(3,4-dichlorophenyl)-3,3-dimethylurea
Molecular Formula
C9H10Cl2N2O
InChI Key
XMTQQYYKAHVGBJ-UHFFFAOYSA-N
SMILES
CN(C)C(=O)NC1=CC=C(Cl)C(Cl)=C1
Appearance (Color)
White to pale cream to pale brown
Assay (HPLC)
≥96.0%
Form
Crystals or powder or crystalline powder or flakes

Description

A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff

Solubility
Soluble in water (0.049g/L), chloroform and methanol. Very slightly soluble in many organic solvents.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text