8-Bromoguanosine hydrate, 97%, Thermo Scientific Chemicals
8-Bromoguanosine hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Bromoguanosine hydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L02992.06
also known as L02992-06
Price (USD)/ Each
62.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
62.20
Add to cart
8-Bromoguanosine hydrate, 97%, Thermo Scientific Chemicals
Catalog numberL02992.06
Price (USD)/ Each
62.20
-
Add to cart
Chemical Identifiers
CAS332359-99-6
IUPAC Name2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H12BrN5O5
InChI KeyASUCSHXLTWZYBA-UMMCILCDSA-N
SMILESNC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Optical Rotation-29.2 ± 0.1? (C=2 in DMSO:Water 1:1)
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream or pink
Water Content (Karl Fischer Titration)≤12.98%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only