Thermo Scientific Chemicals

2-Methylpentane, 99+%, Thermo Scientific Chemicals

Catalog number: L03001.AE
100 mL, Each
Thermo Scientific Chemicals

2-Methylpentane, 99+%, Thermo Scientific Chemicals

Catalog number: L03001.AE
100 mL, Each
Quantity
Catalog number: L03001.AE
also known as L03001-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-83-5
IUPAC Name
2-methylpentane
Molecular Formula
C6H14
InChI Key
AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES
CCCC(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥99.0%
Infrared spectrum
Conforms
Refractive Index
1.3700-1.3730 @ 20?C

Description

2-Methylpentane is employed as a raw material, rubber solvent and vegetable oil extraction solvent. It is also used as a solvent in adhesives. Further, it is used as an intermediate in organic synthesis and finds application in food, preservatives, cosmetics, pharmaceuticals, beverages and flavor enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylpentane is employed as a raw material, rubber solvent and vegetable oil extraction solvent. It is also used as a solvent in adhesives. Further, it is used as an intermediate in organic synthesis and finds application in food, preservatives, cosmetics, pharmaceuticals, beverages and flavor enhancer.

Solubility
Miscible with alcohol, ether, acetone and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text