2-Methyl-8-nitroquinoline, 98%, Thermo Scientific Chemicals
2-Methyl-8-nitroquinoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-8-nitroquinoline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L03006.06
also known as L03006-06
Price (USD)/ Each
94.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
94.50
Add to cart
2-Methyl-8-nitroquinoline, 98%, Thermo Scientific Chemicals
Catalog numberL03006.06
Price (USD)/ Each
94.50
-
Add to cart
Chemical Identifiers
CAS881-07-2
IUPAC Name2-methyl-8-nitroquinoline
Molecular FormulaC10H8N2O2
InChI KeyUHPGVDHXHDPYQP-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream or yellow to orange
FormPowder
Assay (Non-aqueous acid-base Titration)> 97.5%
Assay (GC)> 97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only