Thermo Scientific Chemicals

Ethyl isonicotinate, 98%, Thermo Scientific Chemicals

Catalog number: L03010.22
100 g, Each
Thermo Scientific Chemicals

Ethyl isonicotinate, 98%, Thermo Scientific Chemicals

Catalog number: L03010.22
100 g, Each
Quantity
Catalog number: L03010.22
also known as L03010-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1570-45-2
IUPAC Name
ethyl pyridine-4-carboxylate
Molecular Formula
C8H9NO2
InChI Key
MCRPKBUFXAKDKI-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=NC=C1
Assay (GC)
≥97.5%
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4985-1.5025 @ 20?C

Description

Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text