1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store at 4°C. Air & Light Sensitive. Incompatible with strong bases, strong oxidizing agents, heat and light. Protect from heat.
RUO – Research Use Only
General References:
- Kazutoshi Tanabe. Vibrational frequencies, infrared absorption intensities and energy difference of rotational isomers of 1,2-diiodoethane.Spectrochimica Acta Part A: Molecular Spectroscopy.1974, 30 1901-1914.
- Qingyu Kong; Joonghan Kim; Maciej Lorenc; Tae Kyu Kim; Hyotcherl Ihee and Michael Wulff. Photodissociation Reaction of 1,2-Diiodoethane in Solution: A Theoretical and X-ray Diffraction Study.J. Phys. Chem. A.1974, 109 (45), 10451-10458.
- Source of positive iodine, for example in the formation of 2-iodoindole from lithiated indole-1-carbamate: J. Org. Chem., 57, 2495 (1992), and in the stereoselective formation of (Z)-2-iodoalk-2-enes from ß-oxido phosphonium ylides: J. Chem. Soc., Perkin 1, 1331 (1995).