Thermo Scientific Chemicals

3-Ethyl-3-pentanol, 97%, Thermo Scientific Chemicals

Catalog number: L03025.09
10 g, Each
Thermo Scientific Chemicals

3-Ethyl-3-pentanol, 97%, Thermo Scientific Chemicals

Catalog number: L03025.09
10 g, Each
Quantity
Catalog number: L03025.09
also known as L03025-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
597-49-9
Refractive Index
1.4275-1.4325 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%

Description

3-Ethyl-3-pentanol was used to study the effect of alkanols on the micropolarity and microviscosity of the head group region, in the reverse micelles of AOT-heptane-water by fluorescence probing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-3-pentanol was used to study the effect of alkanols on the micropolarity and microviscosity of the head group region, in the reverse micelles of AOT-heptane-water by fluorescence probing.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store away from acids, acid chlorides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text