Perylene is used as a fluorescent and a chemiluminescent agent in the cytochemistry of membranes. It serves as a blue-emitting dopant material in organic light emitting diodes. It finds application as an organic photoconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perylene is used as a fluorescent and a chemiluminescent agent in the cytochemistry of membranes. It serves as a blue-emitting dopant material in organic light emitting diodes. It finds application as an organic photoconductor.
Solubility
Soluble in chloroform, carbon disulfide and benzene. Slightly soluble in alcohol and ethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Fluorescent and chemiluminescent agent: Anal. Chim. Acta, 97, 21 (1978).
Ali, S.; Bae, J.; Lee, C. H.; Choi, K. H.; Doh, Y. H. Flexible and passive photo sensor based on perylene/graphene composite. Sens. Actuators, B. 2015, 220, 165-173.
Dahan, E.; Sundararajan, P. R. Polymer compatibilized self-assembling perylene derivative. Eur. Polym. J. 2015, 65, 4-14.