(Methylthio)acetonitrile, 99%, Thermo Scientific Chemicals
(Methylthio)acetonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Methylthio)acetonitrile, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number L03066.22
also known as L03066-22
Price (USD)/ Each
246.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
246.00
Add to cart
(Methylthio)acetonitrile, 99%, Thermo Scientific Chemicals
Catalog numberL03066.22
Price (USD)/ Each
246.00
-
Add to cart
Chemical Identifiers
CAS35120-10-6
IUPAC Name2-(methylsulfanyl)acetonitrile
Molecular FormulaC3H5NS
InChI KeyZRIGDBVSVFSVLL-UHFFFAOYSA-N
SMILESCSCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.5%
Appearance (Color)Clear, colorless to yellow
FormLiquid
Refractive Index1.4805-1.4835 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only