Thermo Scientific Chemicals

4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L03071.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L03071.06
5 g, Each
Quantity
Catalog number: L03071.06
also known as L03071-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68837-59-2
IUPAC Name
4-bromo-2-methylbenzoic acid
Molecular Formula
C8H7BrO2
InChI Key
RVCJOGNLYVNRDN-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=CC=C1C(O)=O
Appearance (Color)
White to cream to pale brown or pale pink to pale gray to gray
Form
Crystals or powder or crystalline powder or lumps or granules
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (HPLC)
≥98.0%

Description

A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text