Triphenylcarbenium hexafluorophosphate is a reagent involved in Si-H and Si-Si bond activation at Pt of neutral and cationic silyl Pt bisphosphine complexes. It is used in investigations of interactions with chromium acetylacetonate nitride mononuclear complexes. It is used in double ring-opening polyaddition of spiro orthoesters. It is used as reagent in Synthesis of push-pull tropylium-fused aminoporphyrazine, Preparation of cycloheptatrienyl and tropylium calix[4]arenes, Synthesis of palladacycles containing chiral rhenium half-sandwich complex for use as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triphenylcarbenium hexafluorophosphate is a reagent involved in Si-H and Si-Si bond activation at Pt of neutral and cationic silyl Pt bisphosphine complexes. It is used in investigations of interactions with chromium acetylacetonate nitride mononuclear complexes. It is used in double ring-opening polyaddition of spiro orthoesters. It is used as reagent in Synthesis of “push-pull” tropylium-fused aminoporphyrazine, Preparation of cycloheptatrienyl and tropylium calix[4]arenes, Synthesis of palladacycles containing chiral rhenium half-sandwich complex for use as a catalyst.
Solubility
Soluble in water. (27 g/L) at 25°C.
Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and strong bases. Incompatible with heat and water.
RUO – Research Use Only
Jan Voigt; Thomas Braun. Si-H and Si-Si activation at Pt: synthesis and reactivity of neutral and cationic silyl complexes.Dalton Transactions.2011, 40 (47), 12699-12704.