Thermo Scientific Chemicals

Tetra-n-butoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: L03141.14
25 g, Each
Thermo Scientific Chemicals

Tetra-n-butoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: L03141.14
25 g, Each
Quantity
Catalog number: L03141.14
also known as L03141-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4766-57-8
IUPAC Name
tetrabutyl silicate
Molecular Formula
C16H36O4Si
InChI Key
UQMOLLPKNHFRAC-UHFFFAOYSA-N
SMILES
CCCCO[Si](OCCCC)(OCCCC)OCCCC
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4110-1.4160 @ 20°C (non-U.S. specification)

Description

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text