It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
John S. Peake, et al. Reactions of Silane. I. The Alkylation and Alkoxylation of Silane.J. Am. Chem. Soc.,1952,74(6), 1526-1528.
G. S. Sur, et al. Comparisons among some tetraalkoxysilanes in the hydrolytic precipitation of silica into elastomeric networks.Die Makromolekulare Chemie.,1986,187(12), 2861-2866.