Thermo Scientific Chemicals

Nordihydroguaiaretic acid, 97%, Thermo Scientific Chemicals

Catalog number: L03149.03
1 g, Each
Thermo Scientific Chemicals

Nordihydroguaiaretic acid, 97%, Thermo Scientific Chemicals

Catalog number: L03149.03
1 g, Each
Quantity
Catalog number: L03149.03
also known as L03149-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
500-38-9
IUPAC Name
4-[(2R,3S)-4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
Molecular Formula
C18H22O4
InChI Key
HCZKYJDFEPMADG-TXEJJXNPSA-N
SMILES
C[C@@H](CC1=CC=C(O)C(O)=C1)[C@H](C)CC1=CC=C(O)C(O)=C1
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥96.0% (US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Comment
Material sourced in UK and US
Appearance (Color)
White to cream to pale brown to pale yellow

Description

Nordihydroguaiaretic acid is used in the treatment of multiple diseases, including cardiovascular diseases, neurological disorders and cancers. It is also used as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 100% ethanol, methanol, acetone, dimethylsulfoxide, dimethylformamide, ethanol (5% w/v), glycol, PEG, and slightly soluble in hot water. Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text