Thermo Scientific Chemicals

4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals

Catalog number: L03184.03
1 g, Each
Thermo Scientific Chemicals

4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals

Catalog number: L03184.03
1 g, Each
Quantity
Catalog number: L03184.03
also known as L03184-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6373-50-8
IUPAC Name
1-(2,4-dichlorophenyl)-3-[5-(4-methylphenyl)-1,3,4-thiadiazol-2-yl]urea
Molecular Formula
C16H12Cl2N4OS
InChI Key
YDTTUXLKBBQRHA-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C1=NN=C(NC(=O)NC2=CC=C(Cl)C=C2Cl)S1
Appearance (Color)
White to yellow to orange to cream to pale brown
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
49.0-58.0?C

Description

4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text