Thermo Scientific Chemicals

3,4-Dichlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals

Catalog number: L03226.03
1 g, Each
Thermo Scientific Chemicals

3,4-Dichlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals

Catalog number: L03226.03
1 g, Each
Quantity
Catalog number: L03226.03
also known as L03226-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36480-40-7
IUPAC Name
(3,4-dichlorophenyl)methanethiol
Molecular Formula
C7H6Cl2S
InChI Key
CEGBRSQPRQXALB-UHFFFAOYSA-N
SMILES
SCC1=CC=C(Cl)C(Cl)=C1
Form
Liquid
Refractive Index
1.6045-1.6095 @ 20?C
Assay (GC)
>96.0%
Appearance (Color)
Clear, colorless to pale yellow

Description

Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text