3,4-Dichlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals
3,4-Dichlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dichlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L03226.06
also known as L03226-06
Price (USD)/ Each
175.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
175.00
Add to cart
3,4-Dichlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals
Catalog numberL03226.06
Price (USD)/ Each
175.00
-
Add to cart
Chemical Identifiers
CAS36480-40-7
IUPAC Name(3,4-dichlorophenyl)methanethiol
Molecular FormulaC7H6Cl2S
InChI KeyCEGBRSQPRQXALB-UHFFFAOYSA-N
SMILESSCC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.6045-1.6095 @ 20?C
Assay (GC)>96.0%
Appearance (Color)Clear, colorless to pale yellow
Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
RUO – Research Use Only

General References:

  1. Ruo Li Bai.; Chii Mei Lin.; Teh Yung Liu.; Ernest Hamel. Identification of the cysteine residue of .beta.-tubulin alkylated by the antimitotic agent 2,4-dichlorobenzyl thiocyanate, facilitated by separatio of the protein subunits of tubulin by hydrophobic column chromatography.Biochemistry. 1989, 28 (13),5606-5612 .
  2. Howard E. Katz.; Jerainne Johnson.; Andrew J. Lovinger.; Wenjie Li. Naphthalenetetracarboxylic Diimide-Based n-Channel Transistor Semiconductors: Structural Variation and Thiol-Enhanced Gold Contacts.J. Am. Chem. Soc. 2000, 122 (32),7787-7792 .