Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Slightly soluble in water and ether.
Notes
Moisture Sensitive. Store at 4°C. Protect from moisture.Incompatible materials are Oxidizing agents, Bases, Water.
RUO – Research Use Only
E. A. Semenova; D. Ya. Zhinkin; K. A. Andrianov. Reaction of dialkyldichloro- and alkyldichloro-silanes with methylamine.Inorganic And Analytical Chemistry.1962, 11 1945-1947.
Silylating agent for diols; see Appendix 4.