Thermo Scientific Chemicals

Ethyl 2-methylnicotinate, 97%, Thermo Scientific Chemicals

Catalog number: L03256.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 2-methylnicotinate, 97%, Thermo Scientific Chemicals

Catalog number: L03256.14
25 g, Each
Quantity
Catalog number: L03256.14
also known as L03256-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1721-26-2
IUPAC Name
ethyl 2-methylpyridine-3-carboxylate
Molecular Formula
C9H11NO2
InChI Key
TZORNSWZUZDUCU-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CN=C1C
Appearance (Color)
Clear colorless to yellow to dark yellow to orange or yellow-orange
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5020-1.5080 @ 20°C

Description

Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text