Thermo Scientific Chemicals

Methylsulfonylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: L03280.09
10 g, Each
Thermo Scientific Chemicals

Methylsulfonylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: L03280.09
10 g, Each
Quantity
Catalog number: L03280.09
also known as L03280-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2274-42-2
IUPAC Name
2-methanesulfonylacetonitrile
Molecular Formula
C3H5NO2S
InChI Key
FOTRKCAZUSJCQD-UHFFFAOYSA-N
SMILES
CS(=O)(=O)CC#N
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or lumps
Assay (GC)
≥98.0%
Appearance (Color)
White to cream to brown

Description

(Methylsulfonyl)acetonitrile was used in the synthesis of substituted δ-pyrone ring analogues, 4,9-dihydropyrrolo[2,1-b]quinazolines containing electron withdrawing groups at the 3-position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methylsulfonyl)acetonitrile was used in the synthesis of substituted δ-pyrone ring analogues, 4,9-dihydropyrrolo[2,1-b]quinazolines containing electron withdrawing groups at the 3-position.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text