8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.
Solubility
Soluble in water 0.1-0.5 g/100 mL at 17°C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Doris Kunz. Synthetic routes to N-heterocyclic carbene complexes: pyridine-carbene tautomerizations. Angewandte Chemie (International Edition). 2007, 46, (19), 3405-3408.
C E Scharping; C C Duke; G M Holder; D Larden. The hepatic metabolism of two methylquinolines. Carcinogenesis. 1993, 14, (5), 1041-1047.