Thermo Scientific Chemicals

Thioacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L03305.36
500 g, Each
Thermo Scientific Chemicals

Thioacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L03305.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03305.36
also known as L03305-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
507-09-5
IUPAC Name
ethanethioic S-acid
Molecular Formula
C2H4OS
InChI Key
DUYAAUVXQSMXQP-UHFFFAOYSA-N
SMILES
CC(S)=O
Refractive Index
1.4610-1.4665 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (Argentometric Titration)
≥96.0%
Identification (FTIR)
Conforms

Description

Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

Solubility
Soluble in water. (27g/L) at 16°C.

Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents, metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text