Thermo Scientific Chemicals

Heptanoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: L03315.30
250 g, Each
Thermo Scientific Chemicals

Heptanoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: L03315.30
250 g, Each
Quantity
Catalog number: L03315.30
also known as L03315-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2528-61-2
IUPAC Name
heptanoyl chloride
Molecular Formula
C7H13ClO
InChI Key
UCVODTZQZHMTPN-UHFFFAOYSA-N
SMILES
CCCCCCC(Cl)=O
Form
Liquid
Refractive Index
1.4290-1.4325 @ 20?C
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text