4-Methoxychalcone, 97%, Thermo Scientific Chemicals
4-Methoxychalcone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxychalcone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L03319.14
also known as L03319-14
Price (USD)/ Each
51.40
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
51.40
Add to cart
4-Methoxychalcone, 97%, Thermo Scientific Chemicals
Catalog numberL03319.14
Price (USD)/ Each
51.40
-
Add to cart
Chemical Identifiers
CAS959-33-1
IUPAC Name(2E)-3-(4-methoxyphenyl)-1-phenylprop-2-en-1-one
Molecular FormulaC16H14O2
InChI KeyXUFXKBJMCRJATM-FMIVXFBMSA-N
SMILESCOC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Appearance (Color)Pale yellow to yellow
Melting Point (clear melt)71.0-80.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only