Used as the intermediates of OLED, pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as the intermediates of OLED, pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Naoki Kaneda; Theeradetch Detchprohm; Kazumasa Hiramatsu and Nobuhiko Sawaki. Si-Doping in GaN Grown by Metal-Organic Vapor Phase Epitaxy Using Tetraethylsilane. Japanese Journal of Applied Physics. 2007, 35, (2), 4B.
- Toshiharu Takagi; Hiroshi Teranishi; Chiaki Yokoyama; Shinji Takahashi. Ultrasonic speeds and thermodynamic properties for tetramethylsilane, tetraethylsilane and tetraethoxysilane under high pressures. Thermochimica Acta. 1989, 142, (2), 291-299.