Thermo Scientific Chemicals

Allyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: L03359.22
100 g, Each
Thermo Scientific Chemicals

Allyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: L03359.22
100 g, Each
Quantity
Catalog number: L03359.22
also known as L03359-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1746-13-0
IUPAC Name
(prop-2-en-1-yloxy)benzene
Molecular Formula
C9H10O
InChI Key
POSICDHOUBKJKP-UHFFFAOYSA-N
SMILES
C=CCOC1=CC=CC=C1
Appearance (Color)
Clear, colourless
Form
liquid
Assay (GC)
>98.5%
Refractive Index
1.5200-1.5230

Description

Allyl phenyl ether is an pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl phenyl ether is an pharmaceutical and OLED intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents, acids, bases. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text