Thermo Scientific Chemicals

Crotonaldehyde, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: L03362.AD
50 mL, Each
Thermo Scientific Chemicals

Crotonaldehyde, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: L03362.AD
50 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03362.AD
also known as L03362-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4170-30-3
IUPAC Name
(2Z)-but-2-enal
Molecular Formula
C4H6O
InChI Key
MLUCVPSAIODCQM-IHWYPQMZSA-N
SMILES
CC=C/C=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0% (sum of isomers, UK-sourced material)
Assay from Supplier's CofA
≥98.0% (US-sourced material)
Comment
Material sourced in UK and US

Description

Crotonaldehyde is used as intermediates. It is an excellent prochiral dienophile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crotonaldehyde is used as intermediates. It is an excellent prochiral dienophile.

Solubility
Soluble in water. (150g/L) at 20°C.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with bases, heat, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text