Thermo Scientific Chemicals

4-Methylsalicylic acid, 99%, Thermo Scientific Chemicals

Catalog number: L03367.14
25 g, Each
Thermo Scientific Chemicals

4-Methylsalicylic acid, 99%, Thermo Scientific Chemicals

Catalog number: L03367.14
25 g, Each
Quantity
Catalog number: L03367.14
also known as L03367-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-85-1
IUPAC Name
2-hydroxy-4-methylbenzoic acid
Molecular Formula
C8H8O3
InChI Key
NJESAXZANHETJV-UHFFFAOYSA-N
SMILES
CC1=CC=C(C(O)=O)C(O)=C1
Appearance (Color)
White to cream to yellow to pale orange to gray
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%

Description

4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

Solubility
Soluble in water 10 g/L @ 20˚C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text