Thermo Scientific Chemicals

N-Allylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: L03377.14
25 g, Each
Thermo Scientific Chemicals

N-Allylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: L03377.14
25 g, Each
Quantity
Catalog number: L03377.14
also known as L03377-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-57-9
IUPAC Name
(prop-2-en-1-yl)thiourea
Molecular Formula
C4H8N2S
InChI Key
HTKFORQRBXIQHD-UHFFFAOYSA-N
SMILES
NC(=S)NCC=C
Appearance (Color)
White to pale cream or pale yellow
Comment
Material Sourced in the U.S. and in other countries
Assay from Supplier's CofA
≥97.5%
Melting Point (clear melt)
69.0-78.0°C
Form
Crystals or powder or crystalline powder or granules or chunks

Description

N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.

Solubility
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text