Thermo Scientific Chemicals

m-Toluenesulfonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: L03419.06
5 g, Each
Thermo Scientific Chemicals

m-Toluenesulfonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: L03419.06
5 g, Each
Quantity
Catalog number: L03419.06
also known as L03419-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
312619-56-0
IUPAC Name
3-methylbenzene-1-sulfonic acid hydrate
Molecular Formula
C7H10O4S
InChI Key
GHLYGGIKYFLYTH-UHFFFAOYSA-N
SMILES
O.CC1=CC(=CC=C1)S(O)(=O)=O
Form
Viscous liquid
Water Content (Karl Fischer Titration)
8.0-11.0% (0.8-1.2 waters of hydration)
Appearance (Color)
Pale brown to dark brown
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Refractive Index
1.5250-1.5310 @ 20?C

Description

m-Toluenesulfonic acid monohydrate is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluenesulfonic acid monohydrate is used in chemical research.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from strong bases, moisture. Incompatible materials are oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text