Thermo Scientific Chemicals

Methylsulfonylacetone, 98+%, Thermo Scientific Chemicals

Catalog number: L03421.09
10 g, Each
Thermo Scientific Chemicals

Methylsulfonylacetone, 98+%, Thermo Scientific Chemicals

Catalog number: L03421.09
10 g, Each
Quantity
Catalog number: L03421.09
also known as L03421-09
Price (USD)
Quantity
-
Have Questions?

Description

Methylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text