Sodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Hareendran, S.; Sathishkumar, S.; Abbas, S.; Mackay, A. M.; Rajan, P. A novel composition for the culture of human adipose stem cells which includes complement C3. Cytotechnology 2010, 62 (5), 389-402.
McMahon S. S.; Nikolskaya N.; Choileáin S. N.; Hennessy N.; O’Brien T.; Strappe P. M.; Gorelov A.; Rochev Y. Thermosensitive hydrogel for prolonged delivery of lentiviral vector expressing neurotrophin-3 in vitro. J. Gene. Med. 2011, 13 (11), 591-601.