Thermo Scientific Chemicals

Sodium beta-glycerophosphate pentahydrate, 97%, Thermo Scientific Chemicals

Catalog number: L03425.22
100 g, Each
Thermo Scientific Chemicals

Sodium beta-glycerophosphate pentahydrate, 97%, Thermo Scientific Chemicals

Catalog number: L03425.22
100 g, Each
Quantity
Catalog number: L03425.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13408-09-8
IUPAC Name
[(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
Molecular Formula
C3H19Na2O11P
InChI Key
VWVLGHQKEJESPW-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)
26.8-31.4% (4.4-5.5 waters; Theory 29.43%)
Appearance (Color)
White to pale cream
Form
Crystalline powder
Identification (FTIR)
Conforms

Description

Sodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text