Cyanuric chloride, 98%, Thermo Scientific Chemicals
Cyanuric chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyanuric chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number L03442.18
also known as L03442-18
Price (USD)/ Each
22.65
Online exclusive
25.20 
Save 2.55 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
22.65
Online exclusive
25.20 
Save 2.55 (10%)
Add to cart
Cyanuric chloride, 98%, Thermo Scientific Chemicals
Catalog numberL03442.18
Price (USD)/ Each
22.65
Online exclusive
25.20 
Save 2.55 (10%)
-
Add to cart
Chemical Identifiers
CAS108-77-0
IUPAC Nametrichloro-1,3,5-triazine
Molecular FormulaC3Cl3N3
InChI KeyMGNCLNQXLYJVJD-UHFFFAOYSA-N
SMILESClC1=NC(Cl)=NC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Anticaking agentMay contain 0% to 0.5% Silica as an additive
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
CommentMay contain small amounts of dark particulate matter from the manufacturing process
View more
Reagent for detection of glycine in presence of other amino acids.Cyanuric chloride is a reagent used for the conversion of alcohols and carboxylic acids to alkyl chlorides and acid chlorides respectively. It acts as an intermediate to prepare dyestuffs, optical brighteners, tanning agents, UV-absorbing agent, softening agents and pharmaceuticals. It is also used as a precursor to dyes and crosslinking agents due to the presence of reactive chlorine atoms. It is used as a dehydrating agent as well as reacting with dimethylformamide to give Gold’s reagent, which is a versatile source of aminoalkylations and a precursor to heterocycles. Further, it is considered as an alternative to oxalyl chloride in the Swern oxidation. In addition to this, it is involved in the synthesis of adenosine receptor ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for detection of glycine in presence of other amino acids.Cyanuric chloride is a reagent used for the conversion of alcohols and carboxylic acids to alkyl chlorides and acid chlorides respectively. It acts as an intermediate to prepare dyestuffs, optical brighteners, tanning agents, UV-absorbing agent, softening agents and pharmaceuticals. It is also used as a precursor to dyes and crosslinking agents due to the presence of reactive chlorine atoms. It is used as a dehydrating agent as well as reacting with dimethylformamide to give Gold’s reagent, which is a versatile source of aminoalkylations and a precursor to heterocycles. Further, it is considered as an alternative to oxalyl chloride in the Swern oxidation. In addition to this, it is involved in the synthesis of adenosine receptor ligand.

Solubility
Soluble in acetone, diethyl ether, dioxane, chloroform, carbon tetrachloride and ketones. Slightly soluble in methanol. Insoluble in cold water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids, alcohols, dimethylformamide, amines and dimethyl sulfoxide.
RUO – Research Use Only

General References:

  1. In the presence of triethylamine, carboxylic acids are converted to their acid chlorides, allowing in situ formation of esters, amides and peptides: Tetrahedron Lett., 20, 3037 (1979). Similarly, sulfonic acids are converted to sulfonyl chlorides: Tetrahedron Lett., 44, 1499 (2003). ω-Hydroxy acids are converted to their lactones: Tetrahedron Lett., 21, 1893 (1980). Mild reagent in ß-lactam synthesis: Synthesis, 209 (1981). Carboxylic acids, including N-Boc, -Fmoc and -Cbz amino acids have been converted to alcohols in good yield by activation with cyanuric chloride and N-methylmorpholine (NMM), followed by reduction with aqueous NaBH4: Tetrahedron Lett., 40, 4395 (1999). Hydroxamic acids can also be prepared in a simple one-flask method using hydroxylamine hydrochloride in the presennce of NMM and DMAP: Org. Lett., 5, 2715 (2003).
  2. Effects deoxygenation of diaryl sulfoxides. Alkyl sulfoxides undergo ɑ-chlorination, which can be avoided by using cyanuric fluoride: Synthesis, 221 (1980).
  3. For use, in combination with DMSO, in a mild and efficient alternative to the Swern oxidation of alcohols to aldehydes or ketones, see: J. Org. Chem., 66, 7907 (2001).
  4. Reagent for dehydration of aldoximes: J. Chem. Soc., Chem. Commun., 1226 (1972), and primary carboxamides: Synthesis, 657 (1980) to nitriles. The Vilsmeier-type complex with DMF also converts aldoximes cleanly to nitriles; ketoximes undergo the Beckmann rearrangement at room temperature in high yield: J. Org. Chem., 67, 6272 (2002). The complex converts primary and secondary alcohols to alkyl chlorides in high yield; addition of NaBr affords mainly the alkyl bromide: Org. Lett., 4, 553 (2002); whereas with 4 eq. of LiF, primary alcohols are selectively formylated, providing a mild and convenient method for their protection: J. Org. Chem., 67, 5152 (2002).
  5. For a brief feature on uses of the reagent in synthesis, see: Synlett, 2156 (2006).