Biphenyl-4,4'-dicarbonitrile, 98%, Thermo Scientific Chemicals
Biphenyl-4,4'-dicarbonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Biphenyl-4,4'-dicarbonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L03450.03
also known as L03450-03
Price (USD)/ Each
94.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
94.50
Add to cart
Biphenyl-4,4'-dicarbonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL03450.03
Price (USD)/ Each
94.50
-
Add to cart
Chemical Identifiers
CAS1591-30-6
IUPAC Name[1,1'-biphenyl]-4,4'-dicarbonitrile
Molecular FormulaC14H8N2
InChI KeyKAXYYLCSSXFXKR-UHFFFAOYSA-N
SMILESN#CC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to green to brown
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (HPLC)≥97.5%
Biphenyl-4,4'-dicarbonitrile is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4,4′-dicarbonitrile is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Atsushi Inoue; Kazuya Kitagawa; Hiroshi Shinokubo; Koichiro Oshima. Simple and Efficient TiCl4-Mediated Synthesis of Biaryls via Arylmagnesium Compounds. Tetrahedron. 2000, 56, (49), 9601-9605.
  2. Yacoub Fall; Henri Doucet Dr.; Maurice Santelli Prof. Palladium-Catalysed Direct 3- or 4-Arylation of 2,5-Disubstituted Pyrrole Derivatives: An Economically and Environmentally Attractive Procedure. Polymer Journal. 2009, 2, (2), 153-157.