Thermo Scientific™

n-Butyl acrylate, 98+%, stab. with up to 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L03476.AE
100 mL, Each
Thermo Scientific™

n-Butyl acrylate, 98+%, stab. with up to 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L03476.AE
100 mL, Each
Quantity
Catalog number: L03476.AE
also known as L03476-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-32-2
IUPAC Name
butyl prop-2-enoate
Molecular Formula
C7H12O2
InChI Key
CQEYYJKEWSMYFG-UHFFFAOYSA-N
SMILES
CCCCOC(=O)C=C
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4160-1.4200 @ 20?C
Solution Test
10% in Diethyl ether: clear solution

Description

n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.

Solubility
Miscible with ethanol, ethyl ether and acetone. Slightly miscible with water and carbon tetrachloride.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, catalytic metals, organotin catalysts, amines, peroxides and halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text