Thermo Scientific Chemicals

3,4-Lutidine, 98+%, Thermo Scientific Chemicals

Catalog number: L03489.AP
500 mL, Each
Thermo Scientific Chemicals

3,4-Lutidine, 98+%, Thermo Scientific Chemicals

Catalog number: L03489.AP
500 mL, Each
Quantity
Catalog number: L03489.AP
also known as L03489-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
583-58-4
IUPAC Name
3,4-dimethylpyridine
Molecular Formula
C7H9N
InChI Key
NURQLCJSMXZBPC-UHFFFAOYSA-N
SMILES
CC1=CC=NC=C1C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.5100-1.5140 @ 20?C

Description

3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.

Solubility
Miscible with water, ethanol and ether.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text