Thermo Scientific Chemicals

1,4-Butanediol, 99%, Thermo Scientific Chemicals

Catalog number: L03491.30
250 g, Each
Thermo Scientific Chemicals

1,4-Butanediol, 99%, Thermo Scientific Chemicals

Catalog number: L03491.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03491.30
also known as L03491-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-63-4
IUPAC Name
butane-1,4-diol
Molecular Formula
C4H10O2
InChI Key
WERYXYBDKMZEQL-UHFFFAOYSA-N
SMILES
OCCCCO
Appearance (Color)
Clear colorless
Form
Liquid/low melting solid
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
<0.5%

Description

1,4-Butanediol is commonly used as a solvent in the chemical industry to manufacture gamma-butyrolactone and elastic fibers like spandex. It is used as a cross-linking agent for thermoplastic urethanes, polyester plasticizers, paints and coatings. It undergoes dehydration in the presence of phosphoric acid yielded teterahydrofuran, which is an important solvent used for various applications. It acts an intermediate and is used to manufacture polytetramethylene ether glycol (PTMEG), polybutylene terephthalate (PBT) and polyurethane (PU). It finds application as an industrial cleaner and a glue remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanediol is commonly used as a solvent in the chemical industry to manufacture gamma-butyrolactone and elastic fibers like spandex. It is used as a cross-linking agent for thermoplastic urethanes, polyester plasticizers, paints and coatings. It undergoes dehydration in the presence of phosphoric acid yielded teterahydrofuran, which is an important solvent used for various applications. It acts an intermediate and is used to manufacture polytetramethylene ether glycol (PTMEG), polybutylene terephthalate (PBT) and polyurethane (PU). It finds application as an industrial cleaner and a glue remover.

Solubility
Miscible with water, ethanol, dimethyl sulfoxide and glycol ethers. Slightly miscible with diethyl ether. Immiscible with aliphatic and aromatic hydrocarbons.

Notes
Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text