Thermo Scientific Chemicals

Diethyl sulfide, 96%, Thermo Scientific Chemicals

Catalog number: L03493.22
100 g, Each
Thermo Scientific Chemicals

Diethyl sulfide, 96%, Thermo Scientific Chemicals

Catalog number: L03493.22
100 g, Each
Quantity
Catalog number: L03493.22
also known as L03493-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
352-93-2
IUPAC Name
(ethylsulfanyl)ethane
Molecular Formula
C4H10S
InChI Key
LJSQFQKUNVCTIA-UHFFFAOYSA-N
SMILES
CCSCC
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.4395-1.4455 @ 20?C

Description

Diethyl sulfide is used as a solvent for anhydrous mineral salts and in plating baths for coating metals with gold or silver. It is used as a flavoring agent. It is also used in the addition reaction with platinum dichloride to prepare platinum ethynyl dimers and polymers with pendant ferrocenyl group. Further, it is used in the preparation of triethylsulfoniuim ethylsulfate by reaction with sulfuric acid diethyl ester. In addition to this, it is used in the quantification procedure of volatile sulfur compounds by solid-phase microextraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl sulfide is used as a solvent for anhydrous mineral salts and in plating baths for coating metals with gold or silver. It is used as a flavoring agent. It is also used in the addition reaction with platinum dichloride to prepare platinum ethynyl dimers and polymers with pendant ferrocenyl group. Further, it is used in the preparation of triethylsulfoniuim ethylsulfate by reaction with sulfuric acid diethyl ester. In addition to this, it is used in the quantification procedure of volatile sulfur compounds by solid-phase microextraction.

Solubility
Miscible with alcohol, ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text