Thermo Scientific Chemicals

N,N'-Dibenzylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L03540.14
25 g, Each
Thermo Scientific Chemicals

N,N'-Dibenzylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L03540.14
25 g, Each
Quantity
Catalog number: L03540.14
also known as L03540-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-28-3
IUPAC Name
benzyl[2-(benzylamino)ethyl]amine
Molecular Formula
C16H20N2
InChI Key
JUHORIMYRDESRB-UHFFFAOYSA-N
SMILES
C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0% (non-U.S. specification)
Comment
Purchased in U.S. and in other countries
Assay from Suppliers CofA
≥96.0% (U.S. specification)
Identification (FTIR)
Conforms

Description

N,N'-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text