Thermo Scientific Chemicals

1-Naphthalenemethylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L03541.06
5 g, Each
Thermo Scientific Chemicals

1-Naphthalenemethylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L03541.06
5 g, Each
Quantity
Catalog number: L03541.06
also known as L03541-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-31-0
IUPAC Name
(naphthalen-1-yl)methanaminium
Molecular Formula
C11H12N
InChI Key
NVSYANRBXPURRQ-UHFFFAOYSA-O
SMILES
[NH3+]CC1=C2C=CC=CC2=CC=C1
Appearance (Color)
Clear or slightly hazy, colorless to yellow to orange to red to dark brown
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.6410-1.6450 @ 20°C

Description

1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.

Solubility
Miscible with ethanol, ether and carbon disulfide.

Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text